Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF41 SIRNA Assays ›
Gene Symbol:
ZNF41
Gene Name:
zinc finger protein 41
Gene Aliases:
MRX89
Chromosome Location:
Chr.X: 47444691 - 47483671 on Build GRCh38
UniGene ID:
Hs.496074
Species:
Human
siRNA ID
HSS111540
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF41
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001324139.1
NP_001311068.1
4
454
NM_001324140.1
NP_001311069.1
5
744
NM_001324141.1
NP_001311070.1
4
483
NM_001324142.1
NP_001311071.1
5
760
NM_001324143.1
NP_001311072.1
4
796
NM_001324144.1
NP_001311073.1
5
834
NM_001324145.1
NP_001311074.1
4
499
NM_001324147.1
NP_001311076.1
5
805
NM_001324148.1
NP_001311077.1
5
850
NM_001324149.1
NP_001311078.1
4
523
NM_001324150.1
NP_001311079.1
6
932
NM_001324151.1
NP_001311080.1
5
874
NM_001324152.1
NP_001311081.1
5
920
NM_001324153.1
NP_001311082.1
5
784
NM_001324154.1
NP_001311083.1
4
671
NM_001324155.1
NP_001311084.1
4
695
NM_007130.3
NP_009061.1
5
1147
NM_153380.3
NP_700359.1
5
1020
XM_006724550.3
XP_006724613.1
5
873
XM_006724555.3
XP_006724618.1
5
1168
XM_017029810.1
XP_016885299.1
5
1248
XM_017029811.1
XP_016885300.1
5
1309
XM_017029812.1
XP_016885301.1
5
1230
XM_017029813.1
XP_016885302.1
5
779
XM_017029814.1
XP_016885303.1
5
1219
XM_017029815.1
XP_016885304.1
5
1268
XM_017029816.1
XP_016885305.1
5
707
XM_017029817.1
XP_016885306.1
5
667
GenBank mRNA
AB209520.1
-
1
856
AJ010017.1
-
4
463
AJ010018.1
-
4
487
AJ010019.1
-
4
533
AJ010020.1
-
4
549
AJ010021.1
-
4
573
AK290021.1
-
5
801
AK294858.1
-
4
396
BC015023.1
-
5
1030
3908304
DA567652.1
-
4
393
DA783869.1
-
4
454
X60155.2
-
5
786
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7745c474b-j25zq:80/100.66.77.222:80.
git-commit: c3c9db9cc82507548abdd0ae6a4ade3f81f66292
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.20.0-2024.11.45-1.0