Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286696.1 | NP_001273625.1 | 3 | 729 | ||
NM_001286697.1 | NP_001273626.1 | 4 | 731 | |||
NM_001322260.1 | NP_001309189.1 | 4 | 860 | |||
NM_007135.2 | NP_009066.2 | 3 | 622 | |||
XM_006717279.3 | XP_006717342.1 | 4 | 830 | |||
GenBank mRNA | AK054606.1 | - | 3 | 614 | ||
AK302284.1 | - | 3 | 671 | |||
AK316233.1 | - | 4 | 667 | |||
BC062309.1 | - | 3 | 577 | 3944657 |