Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286696.1 | NP_001273625.1 | 4, 5 | 847 | ||
NM_001286697.1 | NP_001273626.1 | 5, 6 | 849 | |||
NM_001322260.1 | NP_001309189.1 | 5, 6 | 978 | |||
NM_007135.2 | NP_009066.2 | 4, 5 | 740 | |||
XM_006717279.3 | XP_006717342.1 | 5, 6 | 948 | |||
GenBank mRNA | AK054606.1 | - | 4, 5 | 732 | ||
AK302284.1 | - | 4, 5 | 789 | |||
AK316233.1 | - | 5, 6 | 785 | |||
BC062309.1 | - | 4, 5 | 695 | 3944657 |