Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300776.1 | NP_001287705.1 | 4 | 1219 | ||
NM_001300777.1 | NP_001287706.1 | 5 | 1252 | |||
NM_001300778.1 | NP_001287707.1 | 6 | 1434 | |||
NM_003440.3 | NP_003431.2 | 5 | 1367 | |||
XM_005266188.4 | XP_005266245.1 | 4 | 638 | |||
XM_011534833.2 | XP_011533135.1 | 5 | 716 | |||
XM_011534834.2 | XP_011533136.1 | 5 | 4507 | |||
XM_011534835.2 | XP_011533137.1 | 5 | 4504 | |||
XM_011534836.2 | XP_011533138.1 | 4 | 532 | |||
XM_011534837.2 | XP_011533139.1 | 4 | 657 | |||
XM_011534838.2 | XP_011533140.1 | 6 | 729 | |||
XM_011534839.2 | XP_011533141.1 | 6 | 732 | |||
XM_011534840.2 | XP_011533142.1 | 6 | 697 | |||
XM_017019924.1 | XP_016875413.1 | 6 | 749 | |||
XM_017019925.1 | XP_016875414.1 | 6 | 4574 | |||
GenBank mRNA | AK122741.1 | - | 5 | 1368 | ||
AK296572.1 | - | 5 | 523 | |||
AK303240.1 | - | 4 | 540 | |||
AK313145.1 | - | 5 | 638 | |||
BC022291.1 | - | 6 | 626 | 4724560 | ||
BC040561.1 | - | 5 | 674 | 5268377 | ||
U09368.1 | - | 5 | 721 |