Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300776.1 | NP_001287705.1 | 4 | 1092 | ||
NM_001300778.1 | NP_001287707.1 | 6 | 1307 | |||
NM_003440.3 | NP_003431.2 | 5 | 1240 | |||
XM_005266188.4 | XP_005266245.1 | 4 | 511 | |||
XM_011534833.2 | XP_011533135.1 | 5 | 589 | |||
XM_011534834.2 | XP_011533136.1 | 5 | 4380 | |||
XM_011534835.2 | XP_011533137.1 | 5 | 4377 | |||
XM_011534836.2 | XP_011533138.1 | 4 | 405 | |||
XM_011534837.2 | XP_011533139.1 | 4 | 530 | |||
XM_011534838.2 | XP_011533140.1 | 6 | 602 | |||
XM_011534839.2 | XP_011533141.1 | 6 | 605 | |||
XM_011534840.2 | XP_011533142.1 | 6 | 570 | |||
XM_017019924.1 | XP_016875413.1 | 6 | 622 | |||
XM_017019925.1 | XP_016875414.1 | 6 | 4447 | |||
GenBank mRNA | AK122741.1 | - | 5 | 1241 | ||
AK303240.1 | - | 4 | 413 | |||
AK313145.1 | - | 5 | 511 | |||
BC022291.1 | - | 6 | 499 | 4724560 | ||
BC040561.1 | - | 5 | 547 | 5268377 | ||
U09368.1 | - | 5 | 594 |