Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018011.1 | NP_001018011.1 | 2 | 985 | ||
NM_006006.4 | NP_005997.2 | 2 | 1140 | |||
XM_005271658.4 | XP_005271715.1 | 2 | 1272 | |||
XM_017018257.1 | XP_016873746.1 | 2 | 1073 | |||
XM_017018258.1 | XP_016873747.1 | 2 | 1228 | |||
XM_017018259.1 | XP_016873748.1 | 2 | 1580 | |||
GenBank mRNA | AB208916.1 | - | 2 | 1124 | ||
BC026902.1 | - | 2 | 1044 | 4944546 | ||
BC029812.1 | - | 2 | 1122 | 5175961 | ||
BX648973.1 | - | 2 | 1257 | |||
KU178400.1 | - | 1 | 876 | |||
KU178401.1 | - | 1 | 876 | |||
Z19002.1 | - | 1 | 951 |