Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242884.1 | NP_001229813.1 | 13 | 2025 | ||
NM_001287603.1 | NP_001274532.1 | 14 | 2210 | |||
NM_001287604.1 | NP_001274533.1 | 14 | 2206 | |||
NM_001324137.1 | NP_001311066.1 | 13 | 2003 | |||
NM_001324138.1 | NP_001311067.1 | 13 | 2021 | |||
NM_003443.2 | NP_003434.2 | 14 | 2210 | |||
XM_005245986.1 | XP_005246043.1 | 13 | 2021 | |||
XM_005245987.1 | XP_005246044.1 | 14 | 2151 | |||
XM_011542085.2 | XP_011540387.1 | 13 | 1996 | |||
XM_011542086.1 | XP_011540388.1 | 14 | 2177 | |||
XM_011542087.2 | XP_011540389.1 | 13 | 2018 | |||
XM_011542088.1 | XP_011540390.1 | 14 | 2177 | |||
XM_017002242.1 | XP_016857731.1 | 12 | 1991 | |||
GenBank mRNA | AK223618.1 | - | 14 | 2187 | ||
AK226168.1 | - | 14 | 2150 | |||
AK301896.1 | - | 13 | 1972 | |||
AK303823.1 | - | 13 | 1916 | |||
AK315201.1 | - | 14 | 2157 | |||
BC126163.1 | - | 13 | 2073 | 8991879 | ||
BC143965.1 | - | 13 | 2073 | 9052483 | ||
BQ575263.1 | - | 3 | 554 | |||
BU634371.1 | - | 3 | 575 | |||
U20647.1 | - | 2 | 234 | |||
Y09723.1 | - | 14 | 2097 |