Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260486.1 | NP_001247415.1 | 5 | 1742 | ||
NM_001260487.1 | NP_001247416.1 | 5 | 1625 | |||
NM_001260488.1 | NP_001247417.1 | 6 | 1728 | |||
NM_003445.3 | NP_003436.3 | 5 | 1716 | |||
NM_198089.2 | NP_932355.2 | 5 | 1639 | |||
XM_005259215.3 | XP_005259272.1 | 5 | 1786 | |||
XM_011527278.2 | XP_011525580.1 | 6 | 1671 | |||
XM_011527279.2 | XP_011525581.1 | 6 | 2731 | |||
XM_017027248.1 | XP_016882737.1 | 6 | 1763 | |||
GenBank mRNA | AF187986.1 | - | 5 | 1669 | ||
AK022875.1 | - | 5 | 1606 | |||
AK297360.1 | - | 6 | 1704 | |||
AK297384.1 | - | 4 | 1452 | |||
AK314510.1 | - | 5 | 1621 | |||
BC075050.2 | - | 4 | 1510 | 30915579 | ||
BC075051.2 | - | 4 | 1510 | 30915363 | ||
BC126377.1 | - | 4 | 1564 | 8992093 | ||
BC130381.1 | - | 4 | 1564 | 40146412 |
Gene Symbol: LOC101928063 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110727.1 | 3 | 1773 | |||
GenBank mRNA | AK090553.1 | - | 2 | 2101 | ||
AK094825.1 | - | 3 | 1736 |