Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF185 SIRNA Assays ›
Gene Symbol:
ZNF185
Gene Name:
zinc finger protein 185 (LIM domain)
Gene Aliases:
SCELL
Chromosome Location:
Chr.X: 152898132 - 152973481 on Build GRCh38
UniGene ID:
Hs.16622
Species:
Human
siRNA ID
HSS111653
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF185
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178106.1
NP_001171577.1
23
2130
NM_001178107.1
NP_001171578.1
22
2043
NM_001178108.1
NP_001171579.1
22
2037
NM_001178109.1
NP_001171580.1
21
1947
NM_001178110.1
NP_001171581.1
20
1857
NM_001178113.1
NP_001171584.1
14
1587
NM_001178115.1
NP_001171586.1
8
938
NM_007150.3
NP_009081.2
22
2034
XM_005274730.2
XP_005274787.1
24
2237
XM_005274731.2
XP_005274788.1
24
2234
XM_005274735.2
XP_005274792.1
23
2147
XM_005274737.2
XP_005274794.1
23
2141
XM_005274738.2
XP_005274795.1
23
2138
XM_005274740.2
XP_005274797.1
22
2051
XM_005274741.2
XP_005274798.1
22
2051
XM_005274742.2
XP_005274799.1
22
2048
XM_005274744.2
XP_005274801.1
21
1964
XM_005274745.2
XP_005274802.1
21
1961
XM_005274746.3
XP_005274803.1
20
1880
XM_011531194.1
XP_011529496.1
22
2042
XM_011531195.1
XP_011529497.1
23
2039
XM_017029821.1
XP_016885310.1
25
2369
XM_017029822.1
XP_016885311.1
26
2389
XM_017029823.1
XP_016885312.1
25
2366
XM_017029824.1
XP_016885313.1
24
2285
XM_017029825.1
XP_016885314.1
24
2282
XM_017029826.1
XP_016885315.1
24
2279
XM_017029827.1
XP_016885316.1
24
2276
XM_017029828.1
XP_016885317.1
24
2273
XM_017029829.1
XP_016885318.1
23
2192
XM_017029830.1
XP_016885319.1
23
2186
XM_017029831.1
XP_016885320.1
23
2183
XM_017029832.1
XP_016885321.1
23
2174
XM_017029833.1
XP_016885322.1
22
2108
XM_017029834.1
XP_016885323.1
22
2096
XM_017029835.1
XP_016885324.1
22
2057
GenBank mRNA
AK056517.1
-
11
1093
AK095258.1
-
8
910
AK296967.1
-
22
2037
AK301530.1
-
14
1587
AY997296.1
-
22
2023
BX537525.1
-
8
2612
BX640940.1
-
14
1334
DQ508022.1
-
22
1986
DQ508023.1
-
20
1809
DQ508024.1
-
21
1899
DQ508025.1
-
23
2082
DQ508026.1
-
22
1995
Y09538.1
-
14
1315
Back To Top
More Information
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0