Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321685.1 | NP_001308614.1 | 6 | 1201 | ||
NM_013362.3 | NP_037494.2 | 5 | 1036 | |||
XM_005259223.3 | XP_005259280.1 | 2 | 582 | |||
XM_011527285.2 | XP_011525587.1 | 5 | 1056 | |||
XM_011527286.2 | XP_011525588.1 | 5 | 1700 | |||
GenBank mRNA | AF027138.1 | - | 1 | 121 | ||
AF187991.1 | - | 5 | 1010 | |||
AK223477.1 | - | 5 | 955 | |||
AK292437.1 | - | 5 | 1040 | |||
AK302106.1 | - | 4 | 752 | |||
AK313301.1 | - | 4 | 1043 | |||
BC108912.2 | - | 4 | 833 | 40027157 |
Gene Symbol: ZNF155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA164828.1 | - | 1 | 345 |