Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290104.1 | NP_001277033.1 | 14 | 1921 | ||
NM_003460.2 | NP_003451.1 | 14 | 1948 | |||
XM_011545942.2 | XP_011544244.1 | 13 | 1477 | |||
GenBank mRNA | AK310454.1 | - | 14 | 1948 | ||
AK313003.1 | - | 13 | 1490 | |||
BC096304.3 | - | 13 | 1465 | 40007464 | ||
BC096305.3 | - | 13 | 1438 | 40007468 | ||
BC096306.1 | - | 13 | 1465 | 40007471 | ||
BC096307.3 | - | 13 | 1465 | 40007473 | ||
M90366.1 | - | 13 | 1465 |