Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNALI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003462.3 | NP_003453.2 | 1 | 97 | ||
XM_005271172.2 | XP_005271229.1 | 1 | 79 | |||
GenBank mRNA | AK225935.1 | - | 1 | 76 | ||
AK290502.1 | - | 1 | 21 | |||
AK299918.1 | - | 1 | 77 | |||
BC039074.1 | - | 1 | 80 | 5179639 | ||
BC046117.1 | - | 1 | 47 | 6192652 | ||
BX647913.1 | - | 1 | 97 |