Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAPTM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006762.2 | NP_006753.1 | 5 | 570 | ||
XM_011542098.2 | XP_011540400.1 | 3 | 403 | |||
GenBank mRNA | AK292009.1 | - | 5 | 520 | ||
AK297014.1 | - | 5 | 508 | |||
BC106896.1 | - | 5 | 529 | 40030918 | ||
BC106897.1 | - | 5 | 529 | 40030919 | ||
D42042.1 | - | 5 | 500 | |||
U30498.1 | - | 5 | 501 | |||
U51240.1 | - | 5 | 554 |