Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPHS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014625.3 | NP_055440.1 | 5 | 779 | ||
XM_005245483.3 | XP_005245540.1 | 3 | 624 | |||
GenBank mRNA | AJ279254.1 | - | 5 | 760 |