Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008540.1 | NP_001008540.1 | 1 | 940 | ||
NM_003467.2 | NP_003458.1 | 2 | 719 | |||
GenBank mRNA | AF025375.1 | - | 1 | 624 | ||
AF147204.1 | - | 1 | 978 | |||
AF348491.1 | - | 2 | 667 | |||
AK129916.1 | - | 2 | 713 | |||
AK296674.1 | - | 3 | 612 | |||
AK309641.1 | - | 1 | 868 | |||
AK309885.1 | - | 2 | 818 | |||
AK312244.1 | - | 2 | 713 | |||
AY242129.1 | - | 1 | 624 | |||
AY826773.1 | - | 2 | 130 | |||
BC020968.2 | - | 2 | 677 | 3846345 | ||
BT006660.1 | - | 1 | 624 | |||
D10924.1 | - | 2 | 700 | |||
KU245646.1 | - | 2 | 837 | |||
KU245647.1 | - | 2 | 723 | |||
L01639.1 | - | 2 | 649 | |||
L06797.1 | - | 2 | 696 | |||
M99293.1 | - | 2 | 708 | |||
X71635.1 | - | 2 | 691 |