Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008540.1 | NP_001008540.1 | 1 | 1188 | ||
NM_003467.2 | NP_003458.1 | 2 | 967 | |||
GenBank mRNA | AF025375.1 | - | 1 | 872 | ||
AF147204.1 | - | 1 | 1226 | |||
AF348491.1 | - | 2 | 915 | |||
AK129916.1 | - | 2 | 961 | |||
AK296674.1 | - | 3 | 860 | |||
AK309641.1 | - | 1 | 1116 | |||
AK309885.1 | - | 2 | 1066 | |||
AK312244.1 | - | 2 | 961 | |||
AY242129.1 | - | 1 | 872 | |||
BC020968.2 | - | 2 | 925 | 3846345 | ||
BT006660.1 | - | 1 | 872 | |||
D10924.1 | - | 2 | 948 | |||
KU245646.1 | - | 2 | 1085 | |||
KU245647.1 | - | 2 | 971 | |||
L01639.1 | - | 2 | 897 | |||
L06797.1 | - | 2 | 944 | |||
M99293.1 | - | 2 | 956 | |||
X71635.1 | - | 2 | 939 |