Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003469.4 | NP_003460.2 | 2 | 568 | ||
GenBank mRNA | AK312452.1 | - | 2 | 429 | ||
BC022509.1 | - | 2 | 476 | 4820104 | ||
M25756.1 | - | 2 | 401 |