Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006010.5 | NP_006001.4 | 4 | 560 | ||
GenBank mRNA | AK292243.1 | - | 4 | 473 | ||
BC007282.1 | - | 4 | 456 | |||
BC113588.1 | - | 4 | 483 | 8322640 | ||
BC113590.1 | - | 4 | 483 | 8322642 | ||
BE514267.1 | - | 4 | 567 | |||
BT007110.1 | - | 4 | 454 | |||
M83751.1 | - | 4 | 742 |