Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306074.1 | NP_001293003.1 | 4 | 859 | ||
NM_001306075.1 | NP_001293004.1 | 4 | 847 | |||
NM_006527.3 | NP_006518.1 | 5 | 964 | |||
GenBank mRNA | AK091735.1 | - | 4 | 424 | ||
AK093314.1 | - | 5 | 437 | |||
AK094257.1 | - | 5 | 778 | |||
AK300826.1 | - | 4 | 437 | |||
AK315635.1 | - | 5 | 535 | |||
BC014908.1 | - | 5 | 533 | 3908584 | ||
BC015703.1 | - | 5 | 533 | 3907984 | ||
BT007433.1 | - | 5 | 411 | |||
DB031118.1 | - | 5 | 437 | |||
U75679.1 | - | 5 | 526 | |||
Z71188.1 | - | 5 | 515 |