Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005668.5 | NP_005659.1 | 2 | 532 | ||
NM_175052.2 | NP_778222.1 | 2 | 532 | |||
XM_005272078.2 | XP_005272135.1 | 2 | 532 | |||
XM_011543630.1 | XP_011541932.1 | 2 | 532 | |||
GenBank mRNA | AK292872.1 | - | 2 | 484 | ||
BC027866.1 | - | 2 | 461 | |||
BC053657.1 | - | 2 | 513 | 5752337 | ||
BI858673.1 | - | 2 | 528 | |||
BM921020.1 | - | 2 | 532 | |||
DA862968.1 | - | 2 | 532 | |||
L41680.1 | - | 2 | 414 |