Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326606.1 | NP_001313535.1 | 2 | 195 | ||
NM_001326609.1 | NP_001313538.1 | 3 | 401 | |||
NM_001326611.1 | NP_001313540.1 | 2 | 114 | |||
NM_006303.3 | NP_006294.2 | 2 | 315 | |||
XM_005249847.3 | XP_005249904.1 | 2 | 529 | |||
XM_005249848.2 | XP_005249905.1 | 2 | 205 | |||
XM_005249850.3 | XP_005249907.1 | 2 | 126 | |||
XM_006715772.2 | XP_006715835.1 | 2 | 100 | |||
XM_011515519.1 | XP_011513821.1 | 2 | 130 | |||
GenBank mRNA | AK313037.1 | - | 2 | 321 | ||
BC002853.1 | - | 2 | 275 | 3634481 | ||
BC010156.1 | - | 2 | 275 | 3619946 | ||
BC013630.1 | - | 2 | 283 | 3446074 | ||
CX867709.1 | - | 2 | 167 | |||
U24169.1 | - | 2 | 315 |