Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTERF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301134.1 | NP_001288063.1 | 2 | 357 | ||
NM_001301135.1 | NP_001288064.1 | 4 | 517 | |||
NM_006980.4 | NP_008911.1 | 3 | 416 | |||
XM_005250593.3 | XP_005250650.1 | 3 | 407 | |||
XM_006716126.3 | XP_006716189.1 | 3 | 446 | |||
XM_017012620.1 | XP_016868109.1 | 4 | 424 | |||
GenBank mRNA | AK222730.1 | - | 3 | 416 | ||
AK298469.1 | - | 4 | 489 | |||
AK313641.1 | - | 3 | 398 | |||
AL832861.1 | - | 2 | 326 | |||
BC000965.3 | - | 2 | 352 | |||
BC108648.1 | - | 3 | 374 | 6526378 | ||
BC130389.1 | - | 2 | 342 | 40146420 | ||
BC130391.1 | - | 2 | 342 | 40146422 | ||
Y09615.1 | - | 3 | 399 |