Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001269039.2 | NP_001255968.1 | 5 | 1417 | ||
NM_001324336.1 | NP_001311265.1 | 6 | 1512 | |||
NM_001324337.1 | NP_001311266.1 | 7 | 1740 | |||
NM_007373.3 | NP_031399.2 | 6 | 1555 | |||
NR_136749.1 | 5 | 618 | ||||
XM_017016702.1 | XP_016872191.1 | 6 | 1460 | |||
XM_017016703.1 | XP_016872192.1 | 6 | 7277 | |||
XM_017016704.1 | XP_016872193.1 | 6 | 1459 | |||
GenBank mRNA | AB020669.1 | - | 6 | 1483 | ||
AF054828.1 | - | 6 | 1477 | |||
AF068920.1 | - | 5 | 1206 | |||
AK090820.1 | - | 4 | 576 | |||
AK292833.1 | - | 7 | 1681 | |||
AK293271.1 | - | 5 | 557 | |||
BC044752.1 | - | 6 | 1491 | |||
BC050445.1 | - | 5 | 1410 | 6067669 | ||
BX649055.1 | - | 7 | 1323 |