Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001269039.2 | NP_001255968.1 | 7 | 1681 | ||
NM_001324336.1 | NP_001311265.1 | 8 | 1776 | |||
NM_001324337.1 | NP_001311266.1 | 9 | 2004 | |||
NM_007373.3 | NP_031399.2 | 8 | 1819 | |||
NR_136749.1 | 7 | 882 | ||||
XM_017016702.1 | XP_016872191.1 | 8 | 1724 | |||
XM_017016703.1 | XP_016872192.1 | 8 | 7541 | |||
XM_017016704.1 | XP_016872193.1 | 8 | 1723 | |||
GenBank mRNA | AB020669.1 | - | 8 | 1747 | ||
AF054828.1 | - | 8 | 1741 | |||
AF068920.1 | - | 7 | 1470 | |||
AK090820.1 | - | 6 | 840 | |||
AK292833.1 | - | 9 | 1945 | |||
AK293271.1 | - | 7 | 821 | |||
BC044752.1 | - | 8 | 1755 | |||
BC050445.1 | - | 7 | 1674 | 6067669 | ||
BX649055.1 | - | 9 | 1587 |