Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020638.2 | NP_065689.1 | 2 | 425 | ||
GenBank mRNA | AB037973.1 | - | 2 | 279 | ||
AB047858.1 | - | 2 | 411 | |||
AF263537.1 | - | 2 | 425 | |||
AY358323.1 | - | 2 | 312 | |||
BC069333.1 | - | 2 | 353 | 7262193 | ||
BC096713.1 | - | 2 | 318 | 40000232 | ||
BC098147.1 | - | 2 | 318 | 40000231 | ||
BC098252.1 | - | 2 | 318 | 40000230 |