Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297709.1 | NP_001284638.1 | 7 | 525 | ||
NM_001297710.1 | NP_001284639.1 | 6 | 489 | |||
NM_001297711.1 | NP_001284640.1 | 6 | 480 | |||
NM_003480.3 | NP_003471.1 | 8 | 555 | |||
NR_123733.1 | 8 | 555 | ||||
NR_123734.1 | 7 | 525 | ||||
GenBank mRNA | AK124368.1 | - | 6 | 476 | ||
AK290585.1 | - | 8 | 346 | |||
AK299475.1 | - | 7 | 353 | |||
AK315807.1 | - | 8 | 342 | |||
AU280458.1 | - | 8 | 493 | |||
AY339060.1 | - | 6 | 261 | |||
BC005901.1 | - | 8 | 345 | 4247343 | ||
BU929788.1 | - | 7 | 329 | |||
BX396041.2 | - | 6 | 288 | |||
DN996275.1 | - | 8 | 377 | |||
U37283.1 | - | 8 | 477 |