Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005439.2 | NP_005430.1 | 5, 6 | 343 | ||
NR_026581.1 | 5, 6 | 840 | ||||
GenBank mRNA | AF070539.1 | - | 5, 6 | 473 | ||
AK289869.1 | - | 5, 6 | 343 | |||
AK313998.1 | - | 5, 6 | 644 | |||
BC000898.2 | - | 5, 6 | 413 | 2900086 | ||
BC002340.2 | - | 5, 6 | 453 | 2820309 | ||
BT019393.1 | - | 4, 5 | 280 | |||
BT019432.1 | - | 4, 5 | 280 | |||
CR541700.1 | - | 4, 5 | 280 | |||
U57342.1 | - | 5, 6 | 459 |