Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135823.1 | NP_001129295.1 | 2, 3 | 289 | ||
NM_005086.4 | NP_005077.2 | 2, 3 | 472 | |||
XM_011520853.2 | XP_011519155.1 | 2, 3 | 222 | |||
XM_011520854.2 | XP_011519156.1 | 2, 3 | 396 | |||
XM_011520855.2 | XP_011519157.1 | 2, 3 | 138 | |||
GenBank mRNA | AF016028.2 | - | 2, 3 | 372 | ||
AK092925.1 | - | 2, 3 | 285 | |||
AK304667.1 | - | 3, 4 | 366 | |||
AK314353.1 | - | 2, 3 | 461 | |||
AL136756.1 | - | 2, 3 | 456 | |||
BC062299.1 | - | 2, 3 | 452 | 6376868 | ||
BP341300.1 | - | 2, 3 | 472 | |||
DA073350.1 | - | 2, 3 | 289 | |||
X89105.1 | - | 2, 3 | 294 |