Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173466.1 | NP_001166937.1 | 9 | 1031 | ||
NM_015665.5 | NP_056480.1 | 10 | 1130 | |||
XM_011538778.1 | XP_011537080.1 | 10 | 1062 | |||
XM_011538780.1 | XP_011537082.1 | 9 | 963 | |||
GenBank mRNA | AF226048.1 | - | 10 | 1049 | ||
AJ289857.1 | - | 10 | 1057 | |||
AK000833.1 | - | 10 | 1057 | |||
AK094302.1 | - | 8 | 1310 | |||
AK222509.1 | - | 10 | 1057 | |||
AK293342.1 | - | 9 | 988 | |||
AL110160.1 | - | 4 | 341 | |||
AY237818.1 | - | 9 | 999 | |||
BC000659.2 | - | 10 | 1092 | 3347423 | ||
BC018836.2 | - | 10 | 1092 | |||
BT006912.1 | - | 10 | 967 |