Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006011.3 | NP_006002.1 | 5 | 916 | ||
XM_017022642.1 | XP_016878131.1 | 5 | 1079 | |||
XM_017022643.1 | XP_016878132.1 | 4 | 852 | |||
GenBank mRNA | AK313924.1 | - | 5 | 864 | ||
BC069584.1 | - | 5 | 785 | |||
BC096202.3 | - | 5 | 785 | 40004644 | ||
BC096203.3 | - | 5 | 785 | 40004645 | ||
BC096204.3 | - | 5 | 785 | 40004646 | ||
BC096205.3 | - | 4 | 722 | 40004648 | ||
U33551.1 | - | 5 | 761 | |||
U82762.1 | - | 5 | 786 |