Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003486.5 | NP_003477.4 | 3 | 814 | ||
XM_006721286.3 | XP_006721349.2 | 3 | 457 | |||
XM_017023735.1 | XP_016879224.1 | 3 | 819 | |||
XM_017023736.1 | XP_016879225.1 | 2 | 693 | |||
XM_017023737.1 | XP_016879226.1 | 3 | 406 | |||
GenBank mRNA | AB017908.1 | - | 3 | 782 | ||
AB018009.1 | - | 3 | 807 | |||
AB018542.1 | - | 3 | 757 | |||
AF077866.1 | - | 3 | 794 | |||
AF104032.1 | - | 3 | 808 | |||
BC039692.2 | - | 3 | 839 | 5551612 | ||
BC042600.1 | - | 3 | 808 | 4470593 | ||
M80244.1 | - | 3 | 254 |