Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003487.3 | NP_003478.1 | 12 | 1063 | ||
NM_139215.2 | NP_631961.1 | 12 | 1072 | |||
XM_011525315.1 | XP_011523617.1 | 12 | 1042 | |||
XM_017025181.1 | XP_016880670.1 | 10 | 915 | |||
GenBank mRNA | AK304520.1 | - | 13 | 964 | ||
AK307637.1 | - | 11 | 1048 | |||
AK313223.1 | - | 12 | 1029 | |||
AK314194.1 | - | 12 | 1043 | |||
BC046099.1 | - | 12 | 1004 | 6067044 | ||
U51334.1 | - | 12 | 1043 | |||
X98893.1 | - | 12 | 994 |