Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006532.3 | NP_006523.1 | 4, 5 | 550 | ||
XM_011528330.2 | XP_011526632.2 | 4, 5 | 552 | |||
XM_017027335.1 | XP_016882824.1 | 5, 6 | 534 | |||
GenBank mRNA | AB208794.1 | - | 5, 6 | 531 | ||
AK225481.1 | - | 4, 5 | 489 | |||
AK293105.1 | - | 4, 5 | 483 | |||
BC049195.1 | - | 4, 5 | 532 | 5170024 | ||
BI458899.1 | - | 4, 5 | 552 | |||
U16282.1 | - | 4, 5 | 490 |