Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006532.3 | NP_006523.1 | 5, 6 | 822 | ||
XM_011528330.2 | XP_011526632.2 | 5, 6 | 824 | |||
XM_017027335.1 | XP_016882824.1 | 6, 7 | 806 | |||
XM_017027336.1 | XP_016882825.1 | 1, 2 | 558 | |||
GenBank mRNA | AB208794.1 | - | 6, 7 | 803 | ||
AK225481.1 | - | 5, 6 | 761 | |||
AK293105.1 | - | 5, 6 | 755 | |||
BC049195.1 | - | 5, 6 | 804 | 5170024 | ||
U16282.1 | - | 5, 6 | 762 |