Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYMPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004819.2 | NP_004810.2 | 17 | 2552 | ||
XM_005259286.1 | XP_005259343.1 | 17 | 2505 | |||
XM_011527354.1 | XP_011525656.1 | 18 | 2555 | |||
GenBank mRNA | AB209024.1 | - | 17 | 4715 | ||
AB210010.1 | - | 17 | 2397 | |||
AK130602.1 | - | 8 | 691 | |||
AK300252.1 | - | 17 | 2406 | |||
BC006536.2 | - | 16 | 2259 | |||
BC006567.2 | - | 16 | 2259 | |||
BX649165.1 | - | 16 | 3458 | |||
U49240.1 | - | 12 | 2021 | |||
Y10931.1 | - | 17 | 2370 |