Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYMPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004819.2 | NP_004810.2 | 18 | 2606 | ||
XM_005259286.1 | XP_005259343.1 | 18 | 2559 | |||
XM_011527354.1 | XP_011525656.1 | 19 | 2609 | |||
GenBank mRNA | AB209024.1 | - | 18 | 4769 | ||
AB210010.1 | - | 18 | 2451 | |||
AK130602.1 | - | 9 | 745 | |||
AK300252.1 | - | 18 | 2460 | |||
BC006536.2 | - | 17 | 2313 | |||
BC006567.2 | - | 17 | 2313 | |||
BX649165.1 | - | 17 | 3512 | |||
U49240.1 | - | 13 | 2075 | |||
Y10931.1 | - | 18 | 2424 |