Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004651.3 | NP_004642.2 | 10 | 1486 | ||
XM_005272674.3 | XP_005272731.1 | 10 | 1323 | |||
XM_011543988.1 | XP_011542290.1 | 10 | 1280 | |||
GenBank mRNA | AB073597.1 | - | 10 | 1388 | ||
AK055588.1 | - | 9 | 1543 | |||
AK098394.1 | - | 10 | 1390 | |||
AK294635.1 | - | 10 | 1232 | |||
BC000350.2 | - | 10 | 1360 | |||
BC063668.1 | - | 10 | 1368 | |||
BC140849.1 | - | 10 | 1535 | 9021717 | ||
U44839.1 | - | 10 | 1346 |