Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204466.1 | NP_001191395.1 | 7 | 843 | ||
NM_001204467.1 | NP_001191396.1 | 8 | 1074 | |||
NM_001204468.1 | NP_001191397.1 | 8 | 1105 | |||
NM_005676.4 | NP_005667.2 | 8 | 1074 | |||
NM_152856.2 | NP_690595.1 | 7 | 843 | |||
XM_005272677.4 | XP_005272734.1 | 8 | 1127 | |||
XM_005272678.4 | XP_005272735.1 | 7 | 658 | |||
XM_005272679.4 | XP_005272736.1 | 7 | 658 | |||
XM_006724563.1 | XP_006724626.1 | 4 | 291 | |||
XM_017029884.1 | XP_016885373.1 | 7 | 994 | |||
XM_017029885.1 | XP_016885374.1 | 7 | 769 | |||
GenBank mRNA | AK225175.1 | - | 8 | 1033 | ||
AK292758.1 | - | 7 | 794 | |||
AL137421.1 | - | 9 | 963 | |||
BC000681.2 | - | 5 | 299 | |||
BC003089.1 | - | 7 | 782 | 3536058 | ||
BC004181.2 | - | 8 | 1027 | 2961094 | ||
BC008733.2 | - | 8 | 1059 | 3163064 | ||
BC024153.2 | - | 7 | 689 | 3845258 | ||
BX537969.1 | - | 8 | 1056 | |||
D50912.1 | - | 8 | 925 | |||
KU178411.1 | - | 6 | 450 | |||
KU178412.1 | - | 7 | 681 |