Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281463.1 | NP_001268392.1 | 13 | 2283 | ||
NM_006306.3 | NP_006297.2 | 12 | 2137 | |||
GenBank mRNA | AK291921.1 | - | 12 | 2096 | ||
AK301266.1 | - | 9 | 1361 | |||
BC064368.1 | - | 12 | 2068 | |||
BC112127.1 | - | 12 | 2077 | 8327595 | ||
BC171726.1 | - | 13 | 2223 | |||
BX640922.1 | - | 12 | 2180 | |||
D80000.2 | - | 12 | 2009 | |||
S78271.1 | - | 12 | 2042 |