Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TKTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145933.1 | NP_001139405.1 | 2 | 405 | ||
NM_001145934.1 | NP_001139406.1 | 1 | 350 | |||
NM_012253.3 | NP_036385.3 | 2 | 423 | |||
GenBank mRNA | AK292224.1 | - | 2 | 295 | ||
AK292261.1 | - | 2 | 299 | |||
AK302044.1 | - | 2 | 281 | |||
AK302261.1 | - | 2 | 300 | |||
BC025382.2 | - | 2 | 299 | 4825931 | ||
BG718121.1 | - | 2 | 431 | |||
DA505601.1 | - | 1 | 351 | |||
X91817.1 | - | 1 | 189 |