Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AXIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004655.3 | NP_004646.3 | 3 | 1117 | ||
XM_011525319.2 | XP_011523621.1 | 3 | 1328 | |||
XM_011525320.1 | XP_011523622.1 | 3 | 1317 | |||
XM_011525321.2 | XP_011523623.1 | 3 | 1265 | |||
XM_017025192.1 | XP_016880681.1 | 3 | 984 | |||
XM_017025193.1 | XP_016880682.1 | 3 | 1317 | |||
XM_017025194.1 | XP_016880683.1 | 3 | 1096 | |||
GenBank mRNA | AF078165.1 | - | 2 | 917 | ||
AF205888.1 | - | 2 | 934 | |||
AK025718.1 | - | 2 | 139 | |||
AK315303.1 | - | 3 | 1033 | |||
BC101533.1 | - | 2 | 897 | 8069039 | ||
BC143244.1 | - | 2 | 897 | 9051745 | ||
CR933657.1 | - | 3 | 1142 |