Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006768.4 | NP_006759.3 | 6 | 972 | ||
XM_005253944.4 | XP_005254001.1 | 6 | 1072 | |||
XM_017019992.1 | XP_016875481.1 | 5 | 780 | |||
XM_017019993.1 | XP_016875482.1 | 5 | 763 | |||
XM_017019994.1 | XP_016875483.1 | 4 | 638 | |||
XM_017019995.1 | XP_016875484.1 | 4 | 403 | |||
GenBank mRNA | AB208878.1 | - | 6 | 927 | ||
AF035620.1 | - | 6 | 907 | |||
AK290379.1 | - | 6 | 912 | |||
AK299326.1 | - | 3 | 278 | |||
AK308279.1 | - | 6 | 819 | |||
AY332222.1 | - | 6 | 907 | |||
BC136698.1 | - | 6 | 907 | 9020688 | ||
BC136699.1 | - | 6 | 907 | 9020689 |