Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178010.2 | NP_001171481.1 | 12 | 1111 | ||
NM_001178011.2 | NP_001171482.1 | 10 | 877 | |||
NM_003504.4 | NP_003495.1 | 11 | 1015 | |||
XM_005261285.2 | XP_005261342.1 | 13 | 1204 | |||
XM_005261286.3 | XP_005261343.1 | 12 | 1108 | |||
XM_011530415.1 | XP_011528717.1 | 12 | 1108 | |||
XM_011530416.1 | XP_011528718.1 | 11 | 911 | |||
XM_011530417.2 | XP_011528719.1 | 11 | 1171 | |||
XM_011530418.2 | XP_011528720.1 | 11 | 1171 | |||
XM_017028966.1 | XP_016884455.1 | 6 | 490 | |||
XM_017028967.1 | XP_016884456.1 | 3 | 230 | |||
GenBank mRNA | AF053074.1 | - | 11 | 896 | ||
AF062495.1 | - | 11 | 888 | |||
AF081535.1 | - | 11 | 903 | |||
AJ223728.1 | - | 11 | 902 | |||
AK293123.1 | - | 10 | 812 | |||
AK293338.1 | - | 12 | 1008 | |||
AK303274.1 | - | 11 | 935 | |||
AY358971.1 | - | 11 | 938 | |||
BC006232.2 | - | 11 | 933 | 3946493 | ||
BC010022.2 | - | 11 | 918 | 2964592 | ||
BT006792.1 | - | 11 | 872 | |||
CT841513.1 | - | 11 | 876 |