Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FZD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003505.1 | NP_003496.1 | 1 | 2326 | ||
GenBank mRNA | AB017363.1 | - | 1 | 2326 | ||
AF072872.1 | - | 1 | 2316 | |||
AK314996.1 | - | 1 | 2090 | |||
BC051271.1 | - | 1 | 2214 | 4874253 |
Set Membership: |
G-protein coupled receptor |