Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003531.2 | NP_003522.1 | 1 | 15 | ||
GenBank mRNA | BC127610.1 | - | 1 | 15 | 40002514 |
Gene Symbol: HIST1H3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC096128.3 | - | 1 | 15 | 40002508 | |
BC096129.3 | - | 1 | 15 | 40002509 | ||
BC096130.3 | - | 1 | 15 | 40002510 | ||
BC143046.1 | - | 1 | 15 | 40112088 | ||
BC148243.1 | - | 1 | 15 | 40082529 | ||
BC148250.1 | - | 1 | 15 | 40002516 |