Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200029.1 | NP_001186958.1 | 2, 3 | 963 | ||
NM_001200031.1 | NP_001186960.1 | 2, 3 | 411 | |||
NM_003549.3 | NP_003540.2 | 2, 3 | 1177 | |||
GenBank mRNA | AF036035.1 | - | 2, 3 | 1177 | ||
AF040710.1 | - | 2, 3 | 1097 | |||
AF502910.1 | - | 2, 3 | 175 | |||
AF502912.1 | - | 2, 3 | 941 | |||
AK313455.1 | - | 2, 3 | 1088 | |||
BC005896.1 | - | 2, 3 | 1067 | 4104459 | ||
BC012892.1 | - | 1, 2 | 932 | 3860558 |