Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003986.2 | NP_003977.1 | 9 | 1442 | ||
XM_005253159.2 | XP_005253216.1 | 9 | 1690 | |||
XM_005253160.2 | XP_005253217.1 | 9 | 1687 | |||
XM_005253161.2 | XP_005253218.1 | 9 | 1439 | |||
XM_011520402.1 | XP_011518704.1 | 10 | 1850 | |||
XM_017018403.1 | XP_016873892.1 | 8 | 1372 | |||
GenBank mRNA | AF082868.1 | - | 8 | 1140 | ||
AK290108.1 | - | 8 | 1187 | |||
AK313422.1 | - | 8 | 1190 | |||
BC011034.1 | - | 7 | 1130 | 4042355 | ||
CR456812.1 | - | 7 | 1074 |