Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032296.3 | NP_001027467.2 | 9 | 1449 | ||
NM_001286649.1 | NP_001273578.1 | 8 | 1392 | |||
NM_003576.4 | NP_003567.2 | 9 | 1393 | |||
XM_005254079.4 | XP_005254136.1 | 9 | 1270 | |||
XM_017020794.1 | XP_016876283.1 | 9 | 1566 | |||
XM_017020795.1 | XP_016876284.1 | 9 | 1228 | |||
GenBank mRNA | AF024636.1 | - | 9 | 1162 | ||
AF083420.1 | - | 9 | 1265 | |||
AF339826.1 | - | 8 | 870 | |||
AK297583.1 | - | 10 | 1142 | |||
AK299140.1 | - | 8 | 1109 | |||
BC035578.1 | - | 9 | 1397 | 5539375 | ||
BC065378.1 | - | 9 | 1393 | 6598075 | ||
BT019619.1 | - | 9 | 1084 | |||
CR542227.1 | - | 9 | 1084 | |||
EU668350.1 | - | 9 | 1440 |