Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR0B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021969.2 | NP_068804.1 | 2 | 872 | ||
GenBank mRNA | BC030207.1 | - | 2 | 762 | 5222402 | |
HQ692833.1 | - | 2 | 736 |