Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316350.1 | NP_001303279.1 | 5 | 776 | ||
NM_014236.3 | NP_055051.1 | 6 | 959 | |||
XM_005273313.4 | XP_005273370.1 | 6 | 919 | |||
XM_011544303.2 | XP_011542605.1 | 7 | 1008 | |||
XM_011544304.2 | XP_011542606.1 | 6 | 827 | |||
GenBank mRNA | AB209882.1 | - | 6 | 864 | ||
AF043937.1 | - | 6 | 835 | |||
AJ002190.1 | - | 6 | 910 | |||
AK222773.1 | - | 6 | 915 | |||
AK223304.1 | - | 6 | 918 | |||
AK297982.1 | - | 5 | 737 | |||
AK309994.1 | - | 7 | 1005 | |||
AK311386.1 | - | 5 | 609 | |||
AK314181.1 | - | 6 | 922 | |||
BC000450.2 | - | 6 | 806 | 2821393 |